missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gephyrin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12681-1-AP-150UL
This item is not returnable.
View return policy
Description
Stathmin 1 belongs to the stathmin family of genes. It encodes a ubiquitous cytosolic phosphoprotein proposed to function as an intracellular relay integrating regulatory signals of the cellular environment. Stathmin 1 is involved in the regulation of the microtubule filament system by destabilizing microtubules. It prevents assembly and promotes disassembly of microtubules. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
gephyrin | |
Polyclonal | |
Unconjugated | |
GPHN | |
Domain E, GEPH, gephyrin, GPH, GPHN, GPHRYN, KIAA1385, MPT adenylyltransferase, MPT Mo transferase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10243, 268566, 64845 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q03555, Q8BUV3, Q9NQX3 | |
Gphn | |
gephyrin Fusion Protein Ag3370 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |