missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Geranyl acetate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
It is used primarily as a component of perfumes for creams and soaps and as a flavoring ingredient. It is used particularly in rose, lavender and geranium formulations where a sweet fruity or citrus aroma is desired. It is found to have has anti-oxidant, anti-inflammatory and anti-proliferative activities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used primarily as a component of perfumes for creams and soaps and as a flavoring ingredient. It is used particularly in rose, lavender and geranium formulations where a sweet fruity or citrus aroma is desired. It is found to have has anti-oxidant, anti-inflammatory and anti-proliferative activities.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
It is used primarily as a component of perfumes for creams and soaps and as a flavoring ingredient. It is used particularly in rose, lavender and geranium formulations where a sweet fruity or citrus aroma is desired. It is found to have has anti-oxidant, anti-inflammatory and anti-proliferative activities.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.91 |
Boiling Point | 137°C to 139°C (25 mmHg) |
Flash Point | 104°C (219°F) |
Assay Percent Range | 0.98 |
Odor | Pleasant |
Refractive Index | 1.461 |
Quantity | 25 g |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 196.29 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'