missing translation for 'onlineSavingsMsg'
Learn More
Germanium, 99.999%, (trace metal basis), powder, Thermo Scientific Chemicals Product Code.: 10723265

Germanium, 99.999%, (trace metal basis), powder, Thermo Scientific Chemicals

Product Code. 10723265
50 g, Glass bottle
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10723265

Brand: Thermo Scientific Acros 193110500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-56-4
Molecular Formula Ge
Molecular Weight (g/mol) 72.63
MDL Number MFCD00085310
InChI Key GNPVGFCGXDBREM-UHFFFAOYSA-N
Synonym element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powderShow MoreShow Less
PubChem CID 6326954
ChEBI CHEBI:30441
IUPAC Name germanium
SMILES [Ge]

Specifications

Boiling Point 2830.0°C
Melting Point 937.0°C
Color Black to Gray
Physical Form Glistening Crystalline Powder
Quantity 50 g
Assay Percent Range 99.999%
Merck Index 15, 4441
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in hcl and diluted alkali hydroxides
Formula Weight 72.59
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 5.35
Density 5.3500g/mL
Chemical Name or Material Germanium, 99.999%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463795 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number193110500,193110100
Lot NumberA0456252 Certificate TypeCertificate of Analysis Date12/09/2023 Catalog Number193110500,193110010,193110100
Lot NumberA0451685 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number193110500,193110100
Lot NumberA0446638 Certificate TypeCertificate of Analysis Date17/10/2022 Catalog Number193110500,193110010,193110100
Lot NumberA0396310 Certificate TypeCertificate of Analysis Date02/11/2020 Catalog Number193110500,193110010,193110100
compliance-icons
Product Identifier
  • Germanium, powder
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Germanium, 99.999%, (trace metal basis), powder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.