missing translation for 'onlineSavingsMsg'
Learn More
Germanium dioxide, 99.999%, (trace metal basis), electronic grade, Thermo Scientific Chemicals Product Code.: 10635452

Germanium dioxide, 99.999%, (trace metal basis), electronic grade, Thermo Scientific Chemicals

Product Code. 10635452
10 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10635452

Brand: Thermo Scientific Acros 190000100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-53-8
Molecular Formula GeO2
Molecular Weight (g/mol) 104.59
MDL Number MFCD00011030
InChI Key YBMRDBCBODYGJE-UHFFFAOYSA-N
Synonym germanium dioxide, germania, germanic acid, germanium oxide, germanium iv oxide, germanic oxide crystalline, dioxogermanium, germanic oxide, wln: ge o2
PubChem CID 14796
IUPAC Name dioxogermane
SMILES O=[Ge]=O

Specifications

Melting Point 1115.0°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.999%
Linear Formula GeO2
Merck Index 15,4443
Solubility Information Solubility in water: 0.4%. Other solubilities: soluble in about 100 parts boiling water, soluble in acids or solutions of the fixed, alkalies
Formula Weight 104.59
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Density 6.2300g/mL
Chemical Name or Material Germanium dioxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465236 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog Number190002500,190000100,190000500
Lot NumberA0459189 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog Number190002500,190000100,190000500
Lot NumberA0455788 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number190002500,190000100,190000500
Lot NumberA0403165 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number190002500,190000100,190000500
Lot NumberA0414854 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number190002500,190000100,190000500
compliance-icons
Product Identifier
  • Germanium dioxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Germanium dioxide, 99.999%, (trace metal basis), electronic grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.