Learn More
Specac™ Germanium Golden Gate™ Mk ll ATR
Brand: Specac™
Code : Z3
Additional Details : Weight : 5.60000kg
Description
Golden Gate Single Reflection ATR Accessory with Germanium Top-Plate and ZnSe Lenses. Ideal for QA, analysis of hard and soft polymer pellets, forensic sampling, hard samples, corrosive liquids, coated wires and air sensitive samples. Can be used in all FTIR instruments.Specifications
Can be Used in All FTIR Instruments. |