Learn More
Specac™ Germanium Golden Gate™ Mk ll ATR Grooved Anvil Narrow Gauge
Brand:  Specac™ GS10547
Encompass_Preferred
Product Code. 15230108
  • £618.31 / Each
Explore more special offers
Description
Description
Spare Grooved anvil narrow gauge used in the Golden Gate ATR Accessory GS10500. To study the coating on transformer wire the grooved anvils hold the wire exactly in the middle of the diamond.
Specifications
Specifications
Golden Gate ATR Accessory GS10500