missing translation for 'onlineSavingsMsg'
Learn More
Germanium(IV) chloride, 99.9999% (metals basis), Thermo Scientific Chemicals Product Code.: 11306018

Germanium(IV) chloride, 99.9999% (metals basis), Thermo Scientific Chemicals

Product Code. 11306018
25 g
Click to view available options
Quantity:
0.5 g
5 g
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11306018

Brand: Thermo Scientific Alfa Aesar 010509.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a catalyst in the conversion of carbohydrates into 5-hydroxymethylfurfural in ionic liquids, as a reducing agent in combination with triphenylphosphine (TPP) for the reduction of alpha-bromo carboxylic acid derivatives, and in the production of pure germanium. It also finds use as an intermediate for several optical processes. It is one of the most important dopants in silica glass for optical fibers. It is employed in the preparation of germanium dioxide which is used for wide camera lens, microscopy, IR-transparent glasses/windows/lenses, and for the core of fiber-optic lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10038-98-9
Molecular Formula Cl4GeH8
Molecular Weight (g/mol) 222.49
MDL Number MFCD00011029
InChI Key VDHBHZZBEGQJRK-UHFFFAOYSA-N
Synonym germanium tetrachloride, germanium chloride, germane, tetrachloro, germanium iv chloride, germanium, tetrachloro, gecl4, germanium chloride gecl4, unii-ysv1r803c0, wln: .ge..g4, germanium chloride gecl4 8ciShow MoreShow Less
PubChem CID 66226
SMILES Cl.Cl.Cl.Cl.[GeH4]

Specifications

Boiling Point 83.1°C
Melting Point -49.5°C
Color Colorless
Physical Form Liquid
Quantity 25 g
Linear Formula GeCl4
UN Number UN3264
Merck Index 14,4410
Solubility Information Soluble in ether,benzene,chloroform,carbon tetrachloride. Insoluble in hydrochloric acid and sulphuric acid.
IUPAC Name tetrachlorogermane
Formula Weight 214.4
Percent Purity 99.9999%
Odor Acidic
Sensitivity Moisture sensitive
Density 1.874 g/mL
Assay (metals basis)
Chemical Name or Material Germanium(IV) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN13L007 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog Number010509.22,010509.02,010509.14,010509.36,010509.06
Lot NumberN13L010 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog Number010509.22,010509.02,010509.14,010509.36,010509.06
Lot NumberP17G008 Certificate TypeCertificate of Analysis Date27/12/2023 Catalog Number010509.22,010509.02,010509.14,010509.36,010509.06
Lot NumberP02J001 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number010509.22,010509.14,010509.06
Lot NumberY17H007 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010509.22,010509.14,010509.36,010509.06,010517.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Germanium(IV) chloride, 99.9999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.