missing translation for 'onlineSavingsMsg'
Learn More
Germanium(IV) isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11375397

Germanium(IV) isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11375397
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11375397

Brand: Thermo Scientific Alfa Aesar 042673.14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Germanium(IV) isopropoxide is used as precursor to prepare GeO2 organic silanes, which is used to study the application of optical wave guide. It is used in the synthesis of heterotactic polyactide and also involved in the macromolecule mediated synthesis of amorphous Germania.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21154-48-3
Molecular Formula C12H28GeO4
Molecular Weight (g/mol) 308.982
MDL Number MFCD00015339
InChI Key PKLMYPSYVKAPOX-UHFFFAOYSA-N
Synonym germanium iv isopropoxide, tetraisopropoxygermane, germanium tetrapropan-2-olate, tetraisopropoxy-germane, tetraisopropoxygermanium, isopropyl germanate iv, germanium tetraisopropoxideShow MoreShow Less
PubChem CID 16685043
IUPAC Name tetra(propan-2-yloxy)germane
SMILES CC(C)O[Ge](OC(C)C)(OC(C)C)OC(C)C

Specifications

Color Colorless
Density 1.025 g/mL
Boiling Point 109°C (30 mmHg)
Flash Point 68°C (154°F)
Odor Alcohol-like
Linear Formula Ge[OCH(CH3)2]4
Refractive Index 1.414
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 308.94
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Germanium (IV) isopropoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ09K001 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog Number042673.03,042673.14,042673.22,042673.06
Lot NumberW17H022 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number042673.03,042673.14,042673.06
Lot NumberZ08A006 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number042673.03,042673.14,042673.06
Lot NumberF06K06 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number042673.03,042673.14,042673.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Germanium(IV) isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.