missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11307838
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in Phosphors, Transistors and Diodes, and Infrared Transmitting Glass

Used in phosphors, transistors and diodes, and infrared transmitting glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-53-8
Molecular Formula GeO2
Molecular Weight (g/mol) 104.628
MDL Number MFCD00011030
InChI Key YBMRDBCBODYGJE-UHFFFAOYSA-N
Synonym germanium dioxide, germania, germanic acid, germanium oxide, germanium iv oxide, germanic oxide crystalline, dioxogermanium, germanic oxide, wln: ge o2
PubChem CID 14796
IUPAC Name dioxogermane
SMILES O=[Ge]=O

Specifications

Melting Point 1086°C
Color White
Physical Form Sintered Lump
Quantity 50 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information soluble in water; insoluble in HF and HCl.
Formula Weight 104.59
Odor Odorless
Density 6.239
Chemical Name or Material Germanium(IV) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'