Learn More
Germanium(IV) oxide, 99.9999% (metals basis), Thermo Scientific Chemicals
Description
Germanium dioxide is used in algaculture.A mixture is used as an optical material for optical fibers and optical waveguides It is also used as a catalyst in production of polyethylene terephthalate resin and for production of other germanium compounds. It is used as a feedstock for production of some phosphors and semiconductor materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1086°C |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99.9999% (Metals basis) |
Solubility Information | Soluble in water (4.5g/L at 25°C),acid and alkali. Insoluble in Hydrofluoric acid and hydrochloric acid. |
Formula Weight | 104.59 |
Odor | Odorless |
Density | 6.239 |
Chemical Name or Material | Germanium(IV) oxide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M15F028 | Certificate of Analysis | 15/04/2022 | 010510.22,010510.06,010510.14 |
X27H015 | Certificate of Analysis | 12/04/2022 | 010510.22,010510.06,010510.14 |
C27R010 | Certificate of Analysis | 12/11/2021 | 010510.22,010510.06,010510.14 |
J22M26 | Certificate of Analysis | 12/11/2021 | 010510.22,010510.06,010510.14 |
N21A027 | Certificate of Analysis | 12/11/2021 | 010510.22,010510.06,010510.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.