missing translation for 'onlineSavingsMsg'
Learn More
Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11361248

Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11361248
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11361248

Brand: Thermo Scientific Alfa Aesar 010191.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-56-4
Molecular Formula Ge
Molecular Weight (g/mol) 72.63
MDL Number MFCD00085310
InChI Key GNPVGFCGXDBREM-UHFFFAOYSA-N
Synonym element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powderShow MoreShow Less
PubChem CID 6326954
ChEBI CHEBI:30441
IUPAC Name germanium
SMILES [Ge]

Specifications

Quantity 10 g
Percent Purity 99.999%
Odor Odorless
Form Pieces
Assay (metals basis)
Chemical Name or Material Germanium pieces, 3 to 9mm (0.1 to 0.4 in.)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP18K002 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number010191.30,010191.18,010191.09
Lot NumberX17J002 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog Number010191.30,010191.18,010191.09
Lot NumberZ09H006 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010191.30,010191.18,010191.09
Lot NumberJ12R007 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010191.30,010191.18,010191.09
Lot NumberG18N11 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010191.30,010191.18,010191.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.