missing translation for 'onlineSavingsMsg'
Learn More
Germanium tetrachloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10114170

Germanium tetrachloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10114170
100 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10114170

Brand: Thermo Scientific Acros 197151000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10038-98-9
Molecular Formula Cl4GeH8
Molecular Weight (g/mol) 222.49
MDL Number MFCD00011029
InChI Key VDHBHZZBEGQJRK-UHFFFAOYSA-N
Synonym germanium tetrachloride, germanium chloride, germane, tetrachloro, germanium iv chloride, germanium, tetrachloro, gecl4, germanium chloride gecl4, unii-ysv1r803c0, wln: .ge..g4, germanium chloride gecl4 8ciShow MoreShow Less
PubChem CID 66226
SMILES Cl.Cl.Cl.Cl.[GeH4]

Specifications

Boiling Point 82.0°C to 84.0°C
Melting Point -49.5°C
Color Colorless
Physical Form Liquid
Quantity 100 g
Assay Percent Range 99.99%
Linear Formula GeCl4
Merck Index 15, 4444
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in benzene, ether, carbon disulfid and, other organic solvents, insoluble in concentrated hydrochloric acid
IUPAC Name germanium tetrachloride
Formula Weight 214.4
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 1.84
Density 1.8400g/mL
Chemical Name or Material Germanium tetrachloride, 99.99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463474 Certificate TypeCertificate of Analysis Date06/06/2024 Catalog Number197151000,197150250,197150050
Lot NumberA0458805 Certificate TypeCertificate of Analysis Date15/12/2023 Catalog Number197151000,197150250,197150050
Lot NumberA0451061 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number197151000,197150250,197150050
Lot NumberA0444628 Certificate TypeCertificate of Analysis Date29/07/2022 Catalog Number197151000,197150250,197150050
Lot NumberA0438348 Certificate TypeCertificate of Analysis Date20/01/2022 Catalog Number197151000,197150250,197150050
compliance-icons compliance-icons
Product Identifier
  • Germanium tetrachloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Germanium tetrachloride, 99.99%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.