missing translation for 'onlineSavingsMsg'
Learn More

Gestodene, Thermo Scientific™

Product Code. 17362491 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 17362491

Brand: Thermo Scientific Acros 468652500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 60282-87-3
Molecular Formula C21H26O2
Molecular Weight (g/mol) 310.44
MDL Number 00867858
InChI Key SIGSPDASOTUPFS-KQMXEUTGSA-N
Synonym 13beta-Ethyl-17beta-hydroxy-18,19-dinorpregna-4,15-dien-20-yn-3-one
IUPAC Name (1R)-11a-ethyl-1-ethynyl-1-hydroxy-1H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES CCC12CCC3C(CCC4=CC(=O)CCC34)C1C=C[C@@]2(O)C#C

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White to Beige
Melting Point 197.9°C
Merck Index 4425
Quantity 250 mg
Formula Weight 310.43
Chemical Name or Material Gestodene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.