missing translation for 'onlineSavingsMsg'
Learn More

GFAP (Glial Fibrillary Acidic Protein) Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Brand:  Epredia MS-1376-P

276.32 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.


 View more versions of this product

Product Code. 12639686

  • £326.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Glial Fibrillary Acidic Protein (GFAP) is specific to astrocytes (i.e., glial cells) and ependymal cells of the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS. Ab-6 stains astrocytes, glial cells, ependymal cells and their corresponding tumors. Many types of neural tumors such as neuroblastomas, Schwannomas, as well as extra-CNS tumors are not labeled.

Host Species: Mouse

Clone: ASTRO6

Isotype: IgG1

Species Reactivity: Human, Pig, Rat and Chicken. Others not known.

Epitope: Not determined

Immunogen: Glial Fibrillary Acidic Protein

Molecular Weight: 51-52 kDa

Positive Control: IMR-5 cells, Brain or astrocytoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
Specifications

Specifications

GFAP (Glial Fibrillary Acidic Protein)
Monoclonal
200μg/mL
Mouse
1 mL
Neuroscience
Chicken, Human, Porcine, Rat
IgG1
Immunofluorescence
ASTRO6
Unconjugated
Glial Fibrillary Acidic Protein
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers