missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ GFP-4E-BP1
4E-BP1 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for FRAP1 (mTor).
Brand: Thermo Scientific™ PV4759
This item is not returnable.
View return policy
Description
4E-BP1 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for FRAP1 (mTor).
For use in combination with the LanthaScreen™ Tb-anti-4E-BP1 [pThr46] Antibody (Cat. No. PV4757) to evaluate inhibitors in medium to high throughput screening applications. An optimized mTor assay buffer (PV4794) is also available.
Specifications
Biochemical terbium phospho-specific activity | |
Fluorescence | |
Pharma and biopharma, protein assays and analysis, target and lead identification and validation | |
4E-BP1 | |
Kinase Substrate | |
GFP-4E-BP1, 20nmol, 20.02μM Concentration, 45600Da Molecular Weight, Fluorescent Detection Method, GFP (EmGFP Label, Serine/Threonine Kinases Target Enzyme, Used For Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and | |
45,600 Da | |
GFP-4E-BP1 | |
–80°C | |
FRAP1 (mTOR), FRAP1 (mTOR) |
GFP-4E-BP1 protein substrate in 10 mM Tris, pH 7.5, 150 mM NaCl, 10% glycerol, 2 mM DTT Store at -80°C. |
|
Liquid | |
20 nmol | |
Protein-Based Substrate | |
Serine/Threonine Kinases | |
GFP (EmGFP) | |
1 Each | |
Dry Ice | |
mTOR, FRAP1 |
For Research Use Only. Not for use in diagnostic procedures.