Learn More
IBA Lifesciences GFP-Twin-Strep-tag™ Control Protein
Control protein for Strep-Tactin™ and Strep-Tactin™XT applications.
Brand: IBA Lifesciences 2-1007-105
Additional Details : CAS Number : 77-86-1 Weight : 0.00300kg
Description
- Control protein consisting of the green fluorescent protein (GFP) C-terminally tagged with the Twin-Strep-tag™
- Suitable for test purifications, detections, and immobilizations with Strep-Tactin™ or Strep-Tactin™XT
Specifications
1 mg/mL | |
0.5 mg | |
Recombinant | |
Non-sterile | |
Room temperature | |
GFP (Green Fluorescent Protein) | |
Liquid | |
Research Use Only | |
GFP Control Protein |
30 kDa | |
C-terminal GFP-Tag | |
Twin-Strep-tag | |
-25°C to -15°C | |
C-terminal | |
Dissolved in buffer | |
Tagged Control Protein | |
>98% |
For Research Use Only