missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GGTI 298, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
CAAZ peptidomimetic geranylgeranyltransferase I (GGTase I) inhibitor. Strongly inhibits the processing of geranylgeranylated Rap1A with little effect on processing of farnesylated Ha-Ras (IC50 values are 3 and > 10 μM respectively). Causes G0-G1 cell cycle block and apoptosis in A549 cells and inhibits cell invasion and migration in COLO 320CM cells.
Specifications
CAS | 1217457-86-7 |
Molecular Formula | C29H34F3N3O5S |
Synonym | ggti 298, ggti298 trifluoroacetate, ggti 298 tfa salt, ggti-298 tfa salt, ggti 298 tfa, ggti 298 trifluoroacetate salt hydrate, tfa, ggti 298 trifluoroacetate salt hydrate hplc , film, n-4-2-r-amino-3-mercaptopropyl amino-2-naphthylbenzoyl leucine methyl ester trifluoroacetate salt hydrate, n-4-2 r-amino-3-mercaptopropyl amino-2-1-naphthalenyl benzoyl-l-leucine methyl ester trifluoroacetate salt |
InChI Key | WALKWJPZELDSKT-UFABNHQSSA-N |
SMILES | CC(C)CC(C(=O)OC)NC(=O)C1=C(C=C(C=C1)NCC(CS)N)C2=CC=CC3=CC=CC=C32.C(=O)(C(F)(F)F)O |
IUPAC Name | methyl (2S)-2-[[4-[[(2R)-2-amino-3-sulfanylpropyl]amino]-2-naphthalen-1-ylbenzoyl]amino]-4-methylpentanoate;2,2,2-trifluoroacetic acid |
Molecular Weight (g/mol) | 593.662 |
PubChem CID | 16078971 |
Formula Weight | 593.66 |
Percent Purity | >95% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'