Learn More
Brady™ GHS Symbol - Hazardous to Aquatic Environment, Individual Markers on a Roll
Pre-printed labels to identify your chemical product containers.
Brand:  Brady™ 834183
Product Code. 15604837
  • £71.90 / Pack of 250
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products
Specifications
Specifications
PIC 1806-21*21-B7541-RLL-DC
Polyester
Permanent Acrylic
0.099 mm
-40°C to 110°C
Black on White
Gloss
250/Pk.
21 mm
21 mm
Diamond
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ GHS Symbol - Hazardous to Aquatic Environment, Individual Markers on a Roll > W x H: 21 x 21 mm