missing translation for 'onlineSavingsMsg'
Learn More

Brady™ GHS Symbol - Health Hazard, Individual Markers on a Roll

Product Code. 15694827
missing translation for 'orderingAttributeHoverText'
Material:
Polyester
missing translation for 'unitSize'
Pack of 250

Product Code. 15694827

missing translation for 'mfr': Brady™ 834182

Only null left
Add to basket
Edge
Add to basket

Pre-printed labels to identify your chemical product containers.

  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products

Specifications

Alternate Part Number PIC 1805-21*21-B7541-RLL-DC
Finish Gloss
Material Polyester
Quantity 250/Pk.
Adhesive Type Permanent Acrylic
Height (Metric) 21 mm
Thickness 0.099 mm
Width (Metric) 21 mm
Temperature Range (Metric) -40°C to 110°C
Shape Diamond
Color Black on White
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.