missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ 2-Mercaptoethanol

Product Code. 11508916
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11508916

Brand: Gibco™ 21985023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

A potent reducing agent used in cell culture medium to prevent toxic levels of oxygen radicals

  • Mercaptoethanol is not stable in solution. so most protocols require daily supplementation
  • Concentration of 55mM in Dulbecco's Phosphate Buffered Saline (DPBS)
  • Manufactured at a cGMP compliant facility, located in Grand Island, New York, registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 8°C
Shipping conditions: Room temperature
Shelf life: 36 months from date of manufacture
Cell Type Mammalian Cells
Concentration 55 mM
Form Liquid
Product Type BME
pH 6 to 8
Quantity 50 mL
Shipping Condition Room Temperature
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1A
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : 2-Mercaptoethanol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.