missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ 293 F Cells, in CD 293

Faster growth, superior transfection efficiencies and high protein expression levels

Marca:  Gibco™ 11625019

352.56 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Código de producto. 10032812

  • £452.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore más ofertas especiales

For Research Use Only. All usage must comply with product instructions.
 

Alertas:

Handle as potentially biohazardous material under at least Biosafety Level 2 containment. This product contains Dimethyl Sulfoxide (DMSO), a hazardous material. Review the Material Safety Data Sheet before handling.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

Gibco 293-F cells were cloned from the original 293 cell line and adapted to Gibco CD 293 Medium. The 293 cell line is a permanent line established from primary embryonic human kidney and transformed with sheared human adenovirus type 5 DNA. The E1A adenovirus gene is expressed in these cells and participates in transactivation of some viral promoters, allowing these cells to produce very high levels of protein.

  • Quick thaw into suspension culture directly in Gibco CD 293 Medium or Gibco 293 SFM II
  • Faster growth, superior transfection efficiencies and high protein expression levels
  • Documented lineage from a low passage Master Cell Bank
  • Quality and performance testing

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture, Mammalian Expression, Mammalian Expression Cell Lines, Mammalian Protein Production, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Vaccine Production

Información de órdenes

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

1 vial of 7.5 x 106 cells
Storage conditions: liquid nitrogen (vapor phase)
Shipping conditions: frozen
Human
HEK 293 Cells
Dry Ice
293F
SFM (Serum-Free Media)
1.5 mL
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Product Certifications
Ofertas especiales

Ofertas especiales

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado

For Research Use Only. Not for use in diagnostic procedures.