missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Advanced DMEM/F-12
GREENER_CHOICE

Product Code. 11540446
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
500mL
Pack of 10
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11540446

Brand: Gibco™ 12634010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Requires

Supplementation

Advanced DMEM/F-12 (Dulbecco's Modified Eagle Medium/Ham's F-12) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.

Advanced DMEM/F-12 (Dulbecco's Modified Eagle Medium/Ham's F-12) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic DMEM/F-12, serum supplementation can be reduced by 50–90% with no change in growth rate or morphology. Cells successfully cultured in Advanced DMEM/F-12, with no adaptation, include MRC-5, SP2, Vero, WI-38 and Jurkat.

This Advanced DMEM/F-12 is manufactured as follows:

With: Glucose, Non-essential Amino Acids, Sodium Pyruvate, Phenol Red

Without: L-glutamine, HEPES

The complete formulation is available.

Using Advanced DMEM/F-12

Advanced DMEM/F-12 is unique from other media due to addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ II lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride. Advanced DMEM/F-12 requires supplementation with 1–5% Fetal Bovine Serum and 4 mM L-glutamine or GlutaMAX™ supplement. Many cell lines do not require adaptation to this media. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced Advanced DMEM/F-12 uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line MRC-5, SP2, Vero, WI-38 and Jurkat
Classification Animal Origin
Concentration 1 X
Form Liquid
Product Type DMEM (Dulbecco's Modified Eagle Medium)/F-12
Serum Level Reduced Serum
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives High Glucose, Phenol Red, Sodium Pyruvate, Non Essential Amino Acids (NEAA)
Without Additives No Glutamine, No HEPES
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less
compliance-icons
Identifiant produit
  • Methyl benzoate
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
Mention de danger
  • H302-Harmful if swallowed.
Conseils de prudence
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Informations supplémentaires
  • MIXTURE LIST-Contains : Methyl benzoate

For Research Use Only. Not for use in diagnostic procedures.

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis