missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Advanced MEM
GREENER_CHOICE

Product Code. p-4919816
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
5000mL
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11530436

Brand: Gibco™ 12492013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Requires

Supplementation

Advanced MEM (Minimum Essential medium) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.

Advanced MEM (Minimum Essential medium) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic MEM, serum supplementation can be reduced by 50-90% with no change in growth rate or morphology.i Cells successfully cultured in Advanced DMEM / F-12, with no adaptation, include PK-15, MDCK, Vero, WI-38 and Hep-2.

This Advanced MEM is manufactured as follows:

With: Glucose, Non-essential Amino Acids, Sodium Pyruvate, Phenol Red

Without: L-glutamine, HEPES

The complete formulation is available.

Gibco™ Advanced MEM is unique from other media due to addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ I lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride.

Product Use

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Advanced MEM requires supplementation with 1-5% Fetal Bovine Serum and 4mM L-glutamine or GlutaMAX™ supplement. Many cell lines do not require adaptation to this media. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

iPaul Price, Brad Stiles, David Staines, Ethel Evege, Femi Fatunmbi, Kate Wagner, Lori Nestler, David Jayme. Advanced Media as Alternatives to Classical Basal Media. FOCUS, 2003. 25.2:3-6.

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco Advanced MEM is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

iPaul Price, Brad Stiles, David Staines, Ethel Evege, Femi Fatunmbi, Kate Wagner, Lori Nestler, David Jayme. Advanced Media as Alternatives to Classical Basal Media. FOCUS, 2003. 25.2:3-6.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Classification Animal Origin
Concentration 1 X
Form Liquid
Product Type MEM (Minimum Essential Medium)
Serum Level Reduced Serum
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Low Glucose, Phenol Red, Sodium Pyruvate, Non Essential Amino Acids (NEAA)
Without Additives No Glutamine, No HEPES
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 500 mL
Shipping Condition Ambient
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.