missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Agarose (4%)

An autoclaved, high purity, low melting point gel designed for plaque assays and the purification of baculovirus

Brand:  Gibco™ 18300012

Product Code. 11568906

  • £32.83 / 40mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • The 4% Agarose Gel melts at 65 to 70°C and remains fluid at 37°C
  • When diluted with concentrated insect cell culture medium, Gibco™ 4% Agarose produces a gel firm enough to immobilize local infections, yet ductile enough to allow for the efficient removal of plaques for clonal expansion

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2 – 8°C. Protect from light.
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
0.04
Agarose
Autoclaved
40 mL
Room Temperature
Mammalian Cells
Gel
Sterile
Plaque Assay
Not for electrophoresis
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.