missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AmnioMAX™ C-100 Basal Medium
GREENER_CHOICE

Product Code. p-4919818
Click to view available options
Quantity:
90 mL
450 mL
Unit Size:
90mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10538065

Brand: Gibco™ 17001082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Part of kit used to prepare fully-supplemented Gibco AmnioMAX C-100 Complete Medium developed for short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures

Gibco AmnioMAX C-100 Complete Medium is two-part kit for preparation of a complete medium that requires no additional supplementation.

  • Quality and performance testing
  • Every lot of Gibco AmnioMAX C-100 Complete Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance
  • Pooled primary human amniotic fluid samples are cultured for six days in Gibco AmnioMAX C-100 Complete Medium before measuring total number of colonies and total number of mitotic colonies
  • In addition, each lot is tested for sterility, pH and osmolality
  • Unique, optimized formulation
  • Gibco AmnioMAX C-100 Complete Medium contains Fetal Bovine Serum (FBS), gentamicin, and L-glutamine to maximize cell attachment and growth
  • It has enhanced buffering system that provides greater pH stability during culture manipulations
  • Sustainable packaging

Amniotic Fluid and Chorionic Villus Culture, Cell Analysis, Cell Culture, Cell Proliferation, Cell Viability, Proliferation and Function, Clinical, Clinical and Translational Research, Cytogenetics, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco AmnioMAXC-100 Basal Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type C-100 Basal Medium
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine
Quantity 90 mL

Product Intended Use: For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.