Learn More
Gibco™ AmnioMAX™ C-100 Supplement
A part of kit used to prepare fully supplemented Gibco AmnioMAX C-100 Complete Medium developed for short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures
Brand: Gibco™ 12556023
80.33 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
Description
Easy-to-use format: Gibco AmnioMAX C-100 Complete Medium is a two-part kit for preparation of a complete medium that requires no additional supplementation. The complete medium can be stored at 2-8°C for up to ten days
- Quality and performance testing
- Every lot of Gibco AmnioMAX C-100 Complete Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance
- Pooled primary human amniotic fluid samples are cultured for six days in Gibco AmnioMAX C-100 Complete Medium before measuring the total number of colonies and total number of mitotic colonies
- In addition, each lot is tested for sterility, pH and osmolality
- Unique, optimized formulation
- Gibco AmnioMAX C-100 Complete Medium contains Fetal Bovine Serum (FBS), gentamicin, and L-glutamine to maximize cell attachment and growth
- It has enhanced buffering system that provides greater pH stability during culture manipulations
Amniotic Fluid and Chorionic Villus Culture, Cell Analysis, Cell Culture, Cell Proliferation, Cell Viability, Proliferation and Function, Clinical, Clinical and Translational Research, Cytogenetics, Mammalian Cell Culture
Order Info
Shipping Condition: Dry Ice
Compliance
Gibco AmnioMAX C-100 Supplement is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Storage conditions: -5 to -20°C Shipping conditions: Frozen Shelf life: 16 months from date of manufacture. Use complete medium within 7–10 days once opened. |
|
C-100 Supplement | |
Glutamine | |
AmnioMAX™ | |
Dry Ice |
Frozen | |
Sterile-filtered | |
Sustainable packaging | |
75 mL |
In vitro diagnostic