missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Ampicillin, sodium salt, irradiated

Product Code. 10790224
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10790224

Brand: Gibco™ 11593027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Ampicillin is an antibiotic in the broad-spectrum penicillin group

Ampicillin differs from penicillin only by the presence of an amino group, which facilitates penetration through the outer membrane of some gram-negative bacteria. Ampicillin acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that further alter the cell wall.

  • Used as selective antibiotic for resistant bacteria, generally at concentration of 10-25μg/ml for liquid media and 35-50μg/ml for plates
  • Product is provided as powder
  • Made into stock solution at 10mg/ml in water
  • Ampicillin is not used to prevent bacterial contamination of cell cultures due to concerns of mammalian and insect cell toxicity

Cloning, Transformation

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility in Grand Island, NY. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2 to 8°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Concentration 20 to 125 μg/mL
Form Powder
Product Type Antibiotic
For Use With (Application) Bacterial Selection
Quantity 200 mg
Shipping Condition Room Temperature
compliance-icons
Product Identifier
  • AMPICILLIN SODIUM SALT STERILE
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : AMPICILLIN SODIUM SALT

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.