missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Antibiotic-Antimycotic (100X)
GREENER_CHOICE

Product Code. p-4918398
Click to view available options
Quantity:
20 mL
100 mL
20 x 100 mL
Unit Size:
100mL
20mL
Pack of 20
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11580486

Brand: Gibco™ 15240096

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Solution contains penicillin, streptomycin, and Gibco Amphotericin B to prevent cell culture contamination.

Gibco™ Antibiotic-Antimycotic is used to prevent bacterial and fungal contamination. This solution contains 10,000 units/mL of penicillin, 10,000 μg/mL of streptomycin, and 25 μg/mL of Gibco Amphotericin B. The antibiotics penicillin and streptomycin prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. Amphotericin B prevents fungal contamination of cell cultures due to its inhibition of multi-cellular fungus and yeast.

Penicillin was originally purified from the fungus Penicillium and acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that further alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria. Amphotericin B is an antifungal agent that prevents the growth of fungi and yeast by causing an increase in fungal plasma membrane permeability.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: -5 to -20°C
Shipping conditions: Dry ice
Shelf life: 12 months from date of manufacture
Concentration 100 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Antibiotic-Antimycotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
Quantity 20 mL
Shipping Condition Dry Ice
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,2-Diaminopropane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Diaminopropane

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.