Learn More
Gibco™ B-27™ Plus Neuronal Culture System
The B-27 Plus Neuronal Culture System is a next-generation media system that provides the highest rate of in vitro survival of primary rodent and human stem cell-derived neurons.
Brand: Gibco™ A3653401
180.50 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.70000kg
Description
B-27™ Plus Neuronal Culture System is comprised of B-27 Plus Supplement (50X) and Neurobasal Plus Medium, this system is an evolution of the most cited neuronal cell culture products, B-27 Supplement and Neurobasal Medium. It features an optimized formulation, upgraded manufacturing process, and more stringent quality control of raw materials and final product. These improvements enable the highest neuronal cell survival, improved electrophysiological activity, and enhanced functional maturity versus other neuronal cell culture media systems.
The B-27 Plus Neuronal Culture System offers:
- At least 50% higher survival rate of primary rat neurons compared to classic B-27 Supplement and Neurobasal Medium
- Highest survival rate of primary rat, mouse, and human stem cell-derived neurons compared to other commercially available serum-free neuronal media systems
- Improved electrophysiological activity and maturation—more consistent, stable, and synchronized spontaneous activity in primary rodent neurons after 10 days compared to BrainPhys™ Neuronal Medium
- Accelerates neurite outgrowth and increases the number of synaptic-positive puncta
Available as a 50X, 10 mL supplement with 500 mL basal medium, the B-27 Plus Neuronal Culture System can seamlessly replace other neuronal cell culture systems, such as classic B-27 Supplement and Neurobasal Medium. The individual components of the system are also available for purchase separately.
Specifications
• 10 mL B-27 Plus Supplement (50X), store at –5 to –20°C, protect from light • 500 mL Neurobasal Plus Medium, store at 2–8°C, protect from light |
|
Serum-free | |
Liquid | |
Serum-free | |
Ambient |
Neural Cells | |
Mammalian Cell Culture | |
Neuronal Culture System | |
B-27™, Neurobasal™ |
For Research Use Only. Not for use in diagnostic procedures.