missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ B-27™ Plus Supplement (50X)

Product Code. 15717988
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 15717988

Brand: Gibco™ A3582801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

B-27 Plus Supplement is the next-generation of the most cited neuronal cell culture product, B-27 Supplement

B-27 Plus Supplement is a serum-free supplement that provides the highest rate of in vitro survival of primary rodent and human stem cell-derived neurons when used with Neurobasal Plus Medium. B-27 Plus Supplement features an optimized formulation, upgraded manufacturing process, and more stringent quality control of raw materials and final product. These improvements enable the highest rate of neuronal cell survival, improved electrophysiological activity, and enhanced functional maturity versus other neuronal cell culture media systems.


B-27 Plus Supplement, when combined with Neurobasal Plus Medium, offers:

  • At least 50% higher survival rate of primary rat neurons compared to classic B-27 Supplement and Neurobasal Medium
  • Highest survival rate of primary rat, mouse, and human stem cell-derived neurons compared to other commercially available serum-free neuronal media systems
  •  Improved electrophysiological activity and maturation—more consistent, stable, and synchronized spontaneous activity in primary rodent neurons after 10 days compared to BrainPhys™ Neuronal Medium
  •  Accelerates neurite outgrowth and increases the number of synaptic-positive puncta

Available as a 50X, 10 mL supplement, B-27 Plus Supplement can seamlessly replace other neuronal cell culture supplements, such as classic B-27 Supplement. The B-27 Plus Neuronal Culture System is also available for purchase, which is comprised of B-27 Plus Supplement and Neurobasal Plus Medium, offering a convenient and valuable all-in-one ordering solution.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -5 to -20°C. Protect from light.
Cell Type Neural Cells
Classification Serum-free
Concentration 50 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
Product Line B-27
Quantity 10 mL
Shipping Condition Dry Ice
Tested For Endotoxin, Performance, Sterility
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.