missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bovine Serum, heat inactivated, New Zealand origin

Collected from cattle typically 12-24 months of age

Brand:  Gibco™ 26170035

 View more versions of this product

Product Code. 11580636

  • £31.23 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • Sera Category: Secure
  • Origin: New Zealand

Cell Culture, Mammalian Cell Culture

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Liquid
Adult Bovine Serum
Heat-inactivated
Sterile-filtered
Frozen
12 to 24 months
New Zealand
Bottle
100 mL
Sterile-filtered
Gibco™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.