missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bovine Serum, New Zealand origin

US-qualified performance sera for general cell culture with common cell lines

Brand:  Gibco™ 16170086

Code : 50

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11520526

  • £29.21 / 100mL
In Stock
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

  • Sera Category: Secure
  • Origin: New Zealand
  • Collected from cattle of age 12-24 months

Cell Culture, Mammalian Cell Culture

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Liquid
Adult Bovine Serum
Sterile-filtered
Sustainable packaging
Frozen
12 to 24 months
New Zealand
Bottle
100 mL
Sterile-filtered
Gibco™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers