Learn More
Gibco™ Defined Trypsin Inhibitor
Animal origin-free
Brand: Gibco™ R007100
43.12 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Sterile, phosphate-buffered saline solution (1X) containing purified soybean trypsin inhibitor.
Sustainable packaging
Cell Culture, Corneal Epithelial Cell Culture, Keratinocyte Culture, Mammalian Cell Culture, Mammary Epithelial Cell Culture, Primary Cell Culture
Order Info
Shipping Condition: Room Temperature
Specifications
minus 20 degrees C | |
Animal Origin-free | |
Trypsin inhibitor | |
Sustainable packaging | |
Cascade Biologics™ | |
Animal origin-free |
Mammalian Cells | |
1 X | |
Sterile-filtered | |
7.4 | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.