missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CD Hybridoma AGT™ Medium
Animal origin-free, chemically defined, protein-free medium
£101.00 - £5370.00
Specifications
Content And Storage | Storage conditions: 2-8°C, dark and dry Shipping conditions: Gel packs Shelf life: 24 months from date of manufacture |
---|---|
Cell Type | Myeloma Cells, Hybridomas |
Classification | Animal Origin-free, Chemically-defined, Protein-free |
Form | Powder (AGT) |
Product Type | Hybridoma Medium |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10710204
|
Gibco™
12372025 |
1 L |
£101.00
1 L |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11580426
|
Gibco™
12372017 |
10 L |
£890.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10798395
|
Gibco™
12372001 |
100 L |
£5370.00
100 L |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Gibco CD Hybridoma Medium is specifically developed for the ability to support the growth of hybridomas for antibody production.
- Ability to support suspension and stationary hybridomas and myelomas
- Formulated without L-glutamine for stability
- Animal origin-free, protein-free and chemically defined formulation
- Convenient, easy-to-use Advanced Granulation Technology (AGT™) format
Ability to support suspension and stationary hybridomas and myelomas
- Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Gibco CD Hybridoma Medium provides high cell densities and IgG production
- Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate
- Gibco CD Hybridoma Medium requires the addition of 8 mM L-glutamine or GlutaMAX™ supplement before use
- GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format
- Gibco CD Hybridoma Medium is animal origin-free, protein-free and chemically defined, allowing for easier purification of your protein of interest
- Gibco chemically defined media contain no proteins, hydrolysates or components of unknown composition
- The AGT format is an easy-to-use dry powder format. The granules dissolve instantly and pH-adjusts for fast media preparation
- Complete reconstitution instructions are available in the product manual
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Wet ice
Specifications
Storage conditions: 2-8°C, dark and dry Shipping conditions: Gel packs Shelf life: 24 months from date of manufacture |
|
Animal Origin-free, Chemically-defined, Protein-free | |
Hybridoma Medium | |
Non-sterile | |
AGT™, Gibco™ |
Myeloma Cells, Hybridomas | |
Powder (AGT) | |
Serum-free | |
Sodium Pyruvate, Sodium Bicarbonate | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.