missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD Hybridoma Medium
GREENER_CHOICE

Product Code. 10267522
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10267522

Brand: Gibco™ 11279023

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Specifically developed for the growth of hybridomas and myelomas and monoclonal antibody production

CD Hybridoma Medium is an animal origin-free, chemically defined, protein-free medium.

  • Ability to support suspension and stationary hybridomas and myelomas
  • Formulated without L-glutamine for stability
  • Animal origin-free, protein-free, and chemically defined formulation

Ability to support suspension and stationary hybridomas and myelomas

    • Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Gibco CD Hybridoma Medium provides high cell densities and IgG production
    • Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate

Formulated without L-glutamine for stability

    • Gibco CD Hybridoma Medium requires the addition of 8 mM L-glutamine or GlutaMAX™ supplement before use
    • GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format

Animal origin-free, protein-free, and chemically defined formulation

      • Gibco CD Hybridoma Medium is animal origin-free, protein-free, and chemically defined, allowing for easier purification of your protein of interest
      • Gibco chemically defined media contain no proteins, hydrolysates, or components of unknown composition

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Myeloma Cells, Hybridomas
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Liquid
Product Type Hybridoma Medium
Without Additives No Glutamine, No Phenol Red
Product Line Gibco
Quantity 1000 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.