Learn More
Gibco™ Cell Dissociation Buffer, enzyme-free, Hanks' Balanced Salt Solution
Animal origin-free
Brand: Gibco™ 13150016
21.07 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : Weight : 0.15000kg
Description
Gibco Cell Dissociation Buffer is a membrane-filtered, isotonic, and enzyme-free solution of salts, chelating agents, and cell-conditioning agents in calcium-free and magnesium-free Hank's Balanced Salt Solution (HBSS). Gibco Cell Dissociation Buffer is suitable for gentle dissociation of mammalian cells for studies requiring intact cell surface proteins such as ligand binding, flow cytometry, and immunohistochemistry. Gibco Cell Dissociation Buffer works well with lightly adherent cell lines such as HeLa and NIH 3T3, but is not recommended for routine passage of strongly adherent cells.
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room Temperature
Specifications
EDTA | |
HeLa, NIH 3T3 | |
Cell Culture Dissociation Reagent | |
EDTA, Earle's Balanced Salt Solution (EBSS) | |
Room Temperature |
Storage conditions: 15–30°C Shipping conditions: ambient Shelf life: 24 months from date of manufacture |
|
Animal Origin-free | |
Sterile-filtered | |
Room Temperature Stable |
For Research Use Only. Not for use in diagnostic procedures.