Learn More
Gibco™ CF1 Mouse Embryonic Fibroblasts, irradiated
Irradiated Gibco CF1 Mouse Embryonic Fibroblasts (MEFs) are feeder cells that are ideal for supporting healthy undifferentiated human and mouse embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) in culture. Manufactured by MTI-GlobalSt
Brand:  Gibco™ A34181
Additional Details : Weight : 0.07000kg
 View more versions of this product
Product Code. 15943412
  • £58.50 / Pack of 4
In Stock
to see stock.
Description
Description
  • Help save precious time with pre-inactivated, ready-to-use MEFs
  • Culture ESCs and iPSCs with confidence using meticulously tested feeder cells
  • Rely on a widely-used product
Specifications
Specifications
Cryopreserved mouse embryonic fibroblasts
Store in liquid nitrogen.
Feeder Cells (Mouse Embryonic Fibroblast)
Mouse
4 million cells (4 x 106 cells)
Embryonic
Cryopreserved
Pooled
Shipped on dry ice