Learn More
Gibco™ DMEM/F-12, HEPES, no phenol red

DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells.
£20.74 - £192.00
Specifications
Content And Storage | Storage conditions: 2°C to 8°C (protect from light) Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
---|---|
Cell Line | MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts |
Classification | Animal Origin-free |
Concentration | 1X |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11580376
![]() |
Gibco™
11039021 |
500 mL |
£20.74
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11590376
![]() |
Gibco™
11039047 |
10 x 500 mL |
£192.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This DMEM/F-12 is modified as follows:
With: L-glutamine, HEPES
Without: Phenol Red
The complete formulation is available.
Using DMEM/F-12
DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components. DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2°C to 8°C (protect from light) Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Animal Origin-free | |
Liquid | |
Standard Serum Supplementation | |
Sterile-filtered | |
No Phenol Red, No Sodium Pyruvate | |
Gibco™ |
MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts | |
1X | |
DMEM (Dulbecco's Modified Eagle Medium)/F-12 | |
Sterile-filtered | |
High Glucose, Glutamine, HEPES | |
cGMP-compliant under the ISO 13485 standard |
Safety and Handling
missing translation for 'shelfLife' : 12 Months From Date of Manufacture
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.