missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DMEM, high glucose, GlutaMAX™ Supplement, pyruvate
GREENER_CHOICE

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.

£23.85 - £600.00

Specifications

Classification Animal Origin-free
Concentration 1 X
Form Liquid
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
View More Specs

Requires: Supplementation


Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15440544
GREENER_CHOICE
View Documents
Gibco™
10569010
500 mL
£23.85
500mL
Please to purchase this item. Need a web account? Register with us today!
11360872
View Documents
Gibco™
10569069
5 L
£300.00
Each
Please to purchase this item. Need a web account? Register with us today!
11370872
View Documents
Gibco™
10569077
10 L
£600.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:

With: High Glucose, Sodium Pyruvate, GlutaMAX™, Phenol Red

Without: HEPES

The complete formulation is available.

Using DMEM

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Animal Origin-free
Liquid
Sterile-filtered
No HEPES
1 X
Sterile-filtered
High Glucose, GlutaMAX, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months From Date of Manufacture

SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.