missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DMEM, high glucose, no glutamine
GREENER_CHOICE

Product Code. 11500416
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11500416

Brand: Gibco™ 11960044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Requires

Supplementation

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:

With: High Glucose, Phenol Red

Without: L-glutamine, Sodium Pyruvate, HEPES

The complete formulation is available.

Using DMEM

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Compliance

DMEM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. Class II Device U.S.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HeLa, 293, Cos-7, and PC-12
Cell Type Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type DMEM (Dulbecco's Modified Eagle Medium)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives High Glucose, Phenol Red
Without Additives No Glutamine, No HEPES, No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.