Learn More
Gibco™ DMEM, powder, low glucose, pyruvate
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells.
£23.94 - £92.75
Specifications
Content And Storage | Storage conditions: 2-8° C Shipping conditions: Ambient Shelf life: 36 months from date of manufacture |
---|---|
Cell Line | HeLa, 293, Cos-7, and PC-12 |
Cell Type | Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells |
Form | Powder |
Serum Level | Standard Serum Supplementation |
Requires: Supplementation
Description
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This DMEM is modified as follows:
With: Low Glucose, L-glutamine, Sodium Pyruvate, Phenol Red
Without: Sodium Bicarbonate, HEPES
The complete formulation is available.
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2-8° C Shipping conditions: Ambient Shelf life: 36 months from date of manufacture |
|
Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells | |
Standard Serum Supplementation | |
No HEPES, No Sodium Bicarbonate | |
Gibco™ |
HeLa, 293, Cos-7, and PC-12 | |
Powder | |
Low Glucose, Glutamine, Phenol Red, Sodium Pyruvate | |
cGMP-compliant under the ISO 13485 standard | |
Room Temperature |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.