missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS (10X), calcium, magnesium
GREENER_CHOICE

Product Code. 12529069
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 12529069

Brand: Gibco™ 14080055

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

This DPBS is modified as follows:

With: Calcium, Magnesium

Without: Phenol Red

The complete formulation is available.

Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
Concentration 10 X
For Use With (Application) Variety of Cell Culture Applications; Such as Washing Cells Before Dissociation; Transporting Cells or Tissue; Diluting Cells for Counting; and Preparing Reagents
pH 4.4 - 4.7
Physical Form Liquid
Product Line Gibco
Quantity 500 mL
Solution Type Dulbecco's Phosphate Buffered Saline
Sterility Sterile-filtered
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 2900 - 3150 mOsm/kg
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.