missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS, calcium, magnesium
GREENER_CHOICE

Product Code. 10217152
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Osmolality:
270 to 300 mOsm/kg
Unit Size:
1 L
100mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10217152

Brand: Gibco™ 14040117

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.

This DPBS is modified as follows:

With: Calcium, Magnesium

Without: Phenol Red

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
Concentration 1 X
For Use With (Application) Washing Cells Before Dissociation, Transporting Cells or Tissue Samples, Diluting Cells for Counting, and Preparing Reagents
pH 7.0 to 7.2
Physical Form Liquid
Product Line Gibco
Quantity 1000 mL
Solution Type Dulbecco's Phosphate Buffered Saline
Sterility Sterile-filtered
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 270 to 300 mOsm/kg
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.