Learn More
Gibco™ DPBS, calcium, magnesium

Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.
This DPBS is modified as follows:
With: Calcium, Magnesium
Without: Phenol Red
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | DPBS |
| Concentration | 1 X |
| For Use With (Application) | Washing Cells Before Dissociation, Transporting Cells or Tissue Samples, Diluting Cells for Counting, and Preparing Reagents |
| pH | 7.0 to 7.2 |
| Physical Form | Liquid |
| Product Line | Gibco |
| Quantity | 500 mL |
| Solution Type | Dulbecco's Phosphate Buffered Saline |
| Sterility | Sterile-filtered |
| Includes | No Sodium Pyruvate |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.