missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS, no calcium, no magnesium
GREENER_CHOICE

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Brand:  Gibco™ 14190144

15.64 GBP valid until 2025-12-31
Use promo code "24341" to get your promotional price.


 View more versions of this product

Product Code. 15326239

  • £19.18 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

This DPBS is modified as follows:

Without: Calcium, Magnesium, Phenol Red

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Sodium Pyruvate
Clear
1 X
DPBS
Dulbecco's Phosphate Buffered Saline
270 - 300 mOsm/kg
No Phenol Red
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Animal Origin-Free
Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 36 months from date of manufacture
Gibco™
500 mL
Mammalian Cell Culture
7.0 to 7.3
Room Temperature
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3023175 Certificate TypeCertificate of Analysis Date13/02/2025 Catalog Number14190359,14190342,14190144,14190136,14190367
Lot Number3042543 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number14190359,14190342,14190144,14190136,14190367
Lot Number3042529 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number14190359,14190342,14190144,14190136,14190367
Lot Number3062756 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number14190359,14190342,14190144,14190136,14190367
Lot Number2997815 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number14190359,14190342,14190144,14190136,14190367
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ DPBS, no calcium, no magnesium > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.