Learn More
Gibco™ DPBS, no calcium, no magnesium
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
£18.60 - £826.00
Specifications
Classification | Animal Origin-Free |
---|---|
Product Type | DPBS |
Solution Type | Dulbecco's Phosphate Buffered Saline |
For Use With (Application) | Mammalian Cell Culture |
Osmolality | 270 - 300 mOsm/kg |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15326239
|
Gibco™
14190144 |
500 mL |
£18.60
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12559069
|
Gibco™
14190250 |
10 x 500 mL |
£117.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10444402
|
Gibco™
14190136 |
1000 mL |
£19.78
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11544556
|
Gibco™
14190342 |
5 L |
£315.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11554556
|
Gibco™
14190359 |
10 L |
£454.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11340972
|
Gibco™
14190367 |
20 L |
£826.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red
Order Info
Shipping Condition: Room Temperature
Specifications
Animal Origin-Free | |
Dulbecco's Phosphate Buffered Saline | |
270 - 300 mOsm/kg | |
No Phenol Red | |
Liquid |
DPBS | |
Mammalian Cell Culture | |
7.0 to 7.3 | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.