Learn More
Gibco™ Express Five™ SFM
Optimized to support maximum cell growth and recombinant gene expression
Brand: Gibco™ 10486025
99.12 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 1.00000kg
Description
Express FiveSFM is a protein-free, serum-free medium specifically developed to support the long-term suspension growth of High Five (BTI-TN-5B1-4) cells in suspension for use in the Baculovirus Expression Vector System (BEVS). This medium requires the addition of 16 mM L-glutamine before use.
- Superior long-term suspension growth of High Five cells
- Protein-free, serum-free formulation
- Optimized for recombinant protein production
- Scalable in the CelliGen™ bioreactor and WAVE Bioreactor™ System
Cell Culture, High Five™ Cell Culture, Insect Cell Culture
Order Info
Shipping Condition: Room temperature
Compliance
Gibco Express FiveSFM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Storage conditions: 2-8°C. Protect from light Shipping conditions: Ambient Shelf life: 18 months from date of manufacture |
|
Insect Cell | |
Liquid | |
Serum-free | |
Sustainable packaging | |
Room Temperature |
High Five™ | |
Protein-free, Serum-free | |
Insect Cell Serum Free Medium (SFM) | |
No Glutamine | |
Express Five™, Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.