Learn More
Gibco™ Fetal Bovine Serum, certified, heat inactivated, United States

Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
£64.25 - £672.00
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Fetal |
Country of Origin | United States |
Serum Treatment | Heat-inactivated |
Endotoxin Concentration | ≤5 EU/mL |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | |||||
15898937
![]() |
Gibco™
A3840001 |
50 mL | One Shot | N/A | ||||||
15808947
![]() |
Gibco™
A3840002 |
10 x 50 mL | One Shot | N/A | ||||||
11523387
![]() |
Gibco™
10082139 |
100 mL | Bottle | N/A | ||||||
11533387
![]() |
Gibco™
10082147 |
500 mL | Bottle | N/A | ||||||
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Premium (Performance Plus)
- Origin: United States
- Endotoxin level: ≤5 EU/mL
- Hemoglobin level: ≤15 mg/dL
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
United States | |
≤5 EU/mL | |
Certified |
Fetal | |
Heat-inactivated | |
≤15 mg/dL | |
Frozen |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.