missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Embryonic stem-cell FBS, qualified, USDA-approved regions

Embryonic stem-cell FBS, qualified, USDA-approved regions is US certified performance plus sera for broad range of cell types, especially sensitive cell lines.

Brand:  Gibco™ 10439024

 View more versions of this product

Product Code. 11518986

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Specialty
  • Origin: Countries meeting USDA importation requirements
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL

Specially tested for the ability to sustain undifferentiated cellular morphology of embryonic stem cells.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Liquid
Fetal Bovine Serum
Standard (Sterile-filtered)
<50 EU/mL
ES Cell-Qualified
Fetal
Mexico
Bottle
500 mL
Sterile
≤25 mg/dL
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Embryonic stem-cell FBS, qualified, USDA-approved regions > 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.