missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, heat inactivated, USDA-approved regions

Important Update: We've recently introduced Value Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Qualified USDA origin and Brazil (South American) origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

£40.27 - £353.00

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Age Fetal
Country of Origin Countries meeting USDA importation requirements
Form Liquid
Serum Treatment Heat-inactivated
View More Specs

Products 2
Product Code Brand Quantity Format Price Quantity & Availability  
Product Code Brand Quantity Format Price Quantity & Availability  
17296346
View Documents
Gibco™
A3840201
50 mL One Shot This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17256336
View Documents
Gibco™
10438026
500 mL Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Replacing with Fetal Bovine Serum, value
  • Sera application: for cell culture with robust cell lines, good value for basic research
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
  • Origin: countries meeting USDA importation requirements

Gibco serum delivers

  • ISO 13485 certified, processed in FDA registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Countries meeting USDA importation requirements
Heat-inactivated
Triple-filtered, 0.1 μm
Frozen
Fetal
Liquid
Sterile
Qualified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.